Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1GALT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989260
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9260 Supplier Bioss Supplier No. BS9685R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glycosyltransferase that generates the core 1 O-glycan Gal-beta1-3GalNAc-alpha1-Ser/Thr (T antigen), which is a precursor for many extended O-glycans in glycoproteins. Plays a central role in many processes, such as angiogenesis, thrombopoiesis and kidney homeostasis development.

Specifications

Antigen C1GALT1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C1GALT1
Gene Accession No. Q9NS00
Gene Alias 2210410E06Rik; AV284120; B3Gal-T8; Beta-1,3-galactosyltransferase; BOS_4022; C1GALT; C1GALT1; Core 1 beta1,3-galactosyltransferase 1; core 1 beta3-Gal-T; core 1 beta3-Gal-T1; Core 1 O-glycan T-synthase; Core 1 synthase; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1; Core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase (C1galt1); core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 13-galactosyltransferase (C1galt1); Glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; LOC100359047; Plt1; T-syn; T-synthase
Gene Symbols C1GALT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1GALT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56913
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.