Learn More
Invitrogen™ C1orf101 Polyclonal Antibody

Description
C1orf101 Polyclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | C1orf101 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CATSPERE |
| Gene Accession No. | Q5SY80 |
| Gene Alias | C1orf101; Cation channel sperm-associated protein subunit epsilon; CATSPERE; CatSper-epsilon; chromosome 1 open reading frame 101; uncharacterized protein C1orf101 |
| Gene Symbols | CATSPERE |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.