Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112984
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112984 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112984 Supplier Invitrogen™ Supplier No. PA5112984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1orf101 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

Auxiliary component of the CatSper complex, a complex involved in sperm cell hyperactivation. Sperm cell hyperactivation is needed for sperm motility which is essential late in the preparation of sperm for fertilization. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf101
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CATSPERE
Gene Accession No. Q5SY80
Gene Alias C1orf101; Cation channel sperm-associated protein subunit epsilon; CATSPERE; CatSper-epsilon; chromosome 1 open reading frame 101; uncharacterized protein C1orf101
Gene Symbols CATSPERE
Host Species Rabbit
Immunogen Recombinant Human Cation channel sperm-associated protein subunit epsilon protein (601-832AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257044
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.