Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf102 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555849 Supplier Invitrogen™ Supplier No. PA555849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TMSVRTLPLL FLNLGGEMLY ILDQRLRAQN IPGDKARKVL NDIISTMFNR KFMEELFKPQ ELYSKKALRT VYERLAHASI MKLNQASMDK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

May be involved in drug clearance in the placenta. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf102
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OSCP1
Gene Accession No. Q8WVF1
Gene Alias 1810007P19Rik; 5730415O04; 6030436A01Rik; C1orf102; hOSCP1; NOR1; organic solute carrier 1; organic solute carrier partner 1; organic solute carrier protein 1; organic solute transport protein 1; Oscp1; oxidored nitro domain containing protein; oxidored-nitro domain-containing protein; oxidored-nitro domain-containing protein 1; Protein OSCP1; RGD1306596; similar to oxidored-nitro domain-containing protein isoform 1
Gene Symbols OSCP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf102. Recombinant protein control fragment (Product #RP-94096).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127700
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.