Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf103 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983353
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3353 Supplier Bioss Supplier No. BS15001R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The specific function of this protein remains unknown.

Specifications

Antigen C1orf103
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LRIF1
Gene Accession No. Q5T3J3
Gene Alias 2010012G17Rik; 4933421E11Rik; AI450568; C1orf103; HBiX1; HP1-binding protein enriched in inactive X chromosome protein 1; ligand dependent nuclear receptor interacting factor 1; Ligand-dependent nuclear receptor-interacting factor 1; LOC100357914; Lrif1; receptor interacting factor 1; Receptor-interacting factor 1; RGD1306520; RIF1
Gene Symbols LRIF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf103.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55791
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.