Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INAVA Monoclonal Antibody (OTI6A6)
GREENER_CHOICE

Catalog No. PIMA526848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526848 Supplier Invitrogen™ Supplier No. MA526848
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

INAVA Monoclonal Antibody for Western Blot, IHC (P)

Expressed in peripheral macrophages and intestinal myeloid-derived cells, is required for optimal PRR (pattern recognition receptor)-induced signaling, cytokine secretion, and bacterial clearance. Upon stimulation of a broad range of PRRs (pattern recognition receptor) such as NOD2 or TLR2, TLR3, TLR4, TLR5, TLR7 and TLR9, associates with YWHAQ/14-3-3T, which in turn leads to the recruitment and activation of MAP kinases and NF-kappa-B signaling complexes that amplifies PRR-induced downstream signals and cytokine secretion (PubMed:28436939). In the intestine, regulates adherens junction stability by regulating the degradation of CYTH1 and CYTH2, probably acting as substrate cofactor for SCF E3 ubiquitin-protein ligase complexes. Stabilizes adherens junctions by limiting CYTH1-dependent ARF6 activation (PubMed:29420262). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen INAVA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C1orf106
Gene Accession No. Q3KP66
Gene Alias 1700034M08Rik; 4933426C09Rik; 5730559C18Rik; AI586180; C1orf106; chromosome 1 open reading frame 106; D1Mgi54; Inava; Innate immunity activator protein; RGD1311892; RIKEN cDNA 5730559C18 gene; similar to hypothetical protein FLJ10901; uncharacterized protein C1orf106; uncharacterized protein C1orf106 homolog
Gene Symbols C1orf106
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-270 of C1ORF106 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55765
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.