Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INAVA Polyclonal Antibody
C1orf106 Polyclonal Antibody
Supplier: Thermo Scientific PA555635
Description
C1orf106 Polyclonal Antibody for Western Blot

Specifications
INAVA | |
Polyclonal | |
Unconjugated | |
C1orf106 | |
1700034M08Rik; 4933426C09Rik; 5730559C18Rik; AI586180; C1orf106; chromosome 1 open reading frame 106; D1Mgi54; Inava; Innate immunity activator protein; RGD1311892; RIKEN cDNA 5730559C18 gene; similar to hypothetical protein FLJ10901; uncharacterized protein C1orf106; uncharacterized protein C1orf106 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55765 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3KP66 | |
C1orf106 | |
Recombinant protein corresponding to Human C1orf106. Recombinant protein control fragment (Product #RP-94427). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction