Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1orf109 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305934MFV500
Description
C1orf109 Polyclonal antibody specifically detects C1orf109 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
C1orf109 | |
Polyclonal | |
Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
chromosome 1 open reading frame 109, FLJ20508, hypothetical protein LOC54955 | |
Produced in rabbits immunized with E. coli-derived Human C1orf109 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
54955 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction