Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC190 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | C1orf110 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CCDC190 Polyclonal specifically detects CCDC190 in Human samples. It is validated for Western Blot.Specifications
| C1orf110 | |
| Polyclonal | |
| Rabbit | |
| chromosome 1 open reading frame 110, FLJ41579, MGC48998, RP11-331H2.2 | |
| C1ORF110 | |
| IgG | |
| 34 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 339512 | |
| Synthetic peptides corresponding to C1ORF110 The peptide sequence was selected from the middle region of C1ORF110. Peptide sequence SKARNAHYLRHRVPPESERLLSIGEIFGHGESSSSRAGKECENRVPSKFL. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title