Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf111 Monoclonal Antibody (OTI2A9)
GREENER_CHOICE

Catalog No. PIMA527389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527389 Supplier Invitrogen™ Supplier No. MA527389
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C1orf111 Monoclonal Antibody for Western Blot, IHC (P)

Plays a role in spermiogenesis and fertilization. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf111
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPATA46
Gene Accession No. Q5T0L3
Gene Alias 1700015E13Rik; C1orf111; chromosome 1 open reading frame 111; HSD20; RGD1565283; RIKEN cDNA 1700015E13 gene; RP11-565P22.3; similar to novel protein; SPATA46; Spermatogenesis-associated protein 46; uncharacterized protein C1orf111; uncharacterized protein C1orf111 homolog
Gene Symbols SPATA46
Host Species Mouse
Immunogen Full length human recombinant protein of C1ORF111 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284680
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.