Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf111 Monoclonal Antibody (OTI2F3), TrueMAB™, OriGene
SDP

Catalog No. 501683813
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3813 Supplier OriGene Supplier No. TA811094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Play a role in spermiogenesis and fertilization.

Specifications

Antigen C1orf111
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SPATA46
Gene Accession No. Q5T0L3
Gene Alias C1orf111, HSD20
Gene Symbols SPATA46
Host Species Mouse
Immunogen Full length human recombit protein of human C1ORF111 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284680
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.