Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf111 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528542
Description
Recommended positive controls: PC-3, SK-N-SH. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plays a role in spermiogenesis and fertilization. [UniProt].
Specifications
C1orf111 | |
Polyclonal | |
Unconjugated | |
SPATA46 | |
1700015E13Rik; C1orf111; chromosome 1 open reading frame 111; HSD20; RGD1565283; RIKEN cDNA 1700015E13 gene; RP11-565P22.3; similar to novel protein; SPATA46; Spermatogenesis-associated protein 46; uncharacterized protein C1orf111; uncharacterized protein C1orf111 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q5T0L3 | |
SPATA46 | |
Recombinant fragment corresponding to a region within amino acids 1 and 249 of Human C1orf111. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction