Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf123 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113398
Description
C1orf123 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| C1orf123 | |
| Polyclonal | |
| Unconjugated | |
| C1orf123 | |
| C1orf123; chromosome 1 open reading frame 123; CXXC motif containing zinc binding protein; CZIB; UPF0587 protein C1orf123 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 54987 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9NWV4 | |
| C1orf123 | |
| Recombinant Human UPF0587 protein C1orf123 protein (1-160AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction