Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1orf127 Polyclonal Antibody

C1orf127 Polyclonal Antibody
Supplier: Thermo Scientific PA555672
Description
C1orf127 Polyclonal Antibody for IHC

Specifications
C1orf127 | |
Polyclonal | |
Unconjugated | |
C1orf127 | |
C1orf127; chromosome 1 open reading frame 127; uncharacterized protein C1orf127 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
148345 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N9H9 | |
C1orf127 | |
Recombinant protein corresponding to Human C1orf127. Recombinant protein control fragment (Product #RP-94465). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction