Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf127 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546338
Description
Peptide sequence: LVELEYPFQA GRGASLQQEL TEPTLALSAE SHRPPELQDS VEGLSERPSR Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
| C1orf127 | |
| Polyclonal | |
| Unconjugated | |
| C1orf127 | |
| C1orf127; chromosome 1 open reading frame 127; uncharacterized protein C1orf127 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 148345 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N9H9 | |
| C1orf127 | |
| Synthetic peptide directed towards the C-terminal of human C1orf127 (aa 607-656). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction