Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf144 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555018 Supplier Invitrogen™ Supplier No. PA555018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNSTSRPTLP VKSLAQREAE YAEARKRILG SASPEEEQEK PILDRPTRIS QPEDSRQPNN VIRQPLGPDG SQGFKQR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf144
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SZRD1
Gene Accession No. Q7Z422
Gene Alias 1110022I03; C1orf144; C2H1orf144; D4Ertd22e; DKFZp566C0424; putative MAPK activating protein PM20,PM21; putative MAPK-activating protein PM18/PM20/PM22; RGD1560286; similar to putative MAPK activating protein PM20,PM21; SUZ domain-containing protein 1; SUZ RNA binding domain containing 1; SZRD1; UPF0485 protein C1orf144; UPF0485 protein C1orf144 homolog
Gene Symbols SZRD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf144. Recombinant protein control fragment (Product #RP-93996).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26099
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.