Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf144 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9356 Supplier Bioss Supplier No. BS9799R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen C1orf144
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SZRD1
Gene Accession No. Q7Z422
Gene Alias 1110022I03; BOS_2341; C1orf144; C2H1orf144; D4Ertd22e; DKFZp566C0424; LOC100345981; putative MAPK activating protein PM20,PM21; putative MAPK activating protein PM20,PM21-like; Putative MAPK-activating protein PM18/PM20/PM22; RGD1560286; SUZ domain-containing protein 1; SUZ RNA binding domain containing 1; SZRD1; UPF0485 protein C1orf144; UPF0485 protein C1orf144 homolog
Gene Symbols SZRD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf144.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26099
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.