Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1orf146 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP242820
Description
Specifically detects C1orf146 in Human samples, and it is validated for Western Blotting
C1orf146 Polyclonal specifically detects C1orf146 in Human samples. It is validated for Western Blot.Specifications
C1orf146 | |
Polyclonal | |
Western Blot 1:500-1:3000 | |
SCRE, SPO16 | |
Recombinant protein encompassing a sequence within the center region of human C1orf146. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Rabbit | |
Affinity Purified | |
RUO | |
388649 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction