Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf146 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145065
Description
Immunogen sequence: VNAINLMCTI AKTTSKPYID SICYRMITAK AYIIEQSPVW KTLQKIKLNS DSVNP Antigen sequence identity: Mouse 85%; Rat 82%.

Specifications
C1orf146 | |
Polyclonal | |
Unconjugated | |
C1orf146 | |
C1orf146; chromosome 1 open reading frame 146; RP11-163M2.4; uncharacterized protein C1orf146 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
388649 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VVC0 | |
C1orf146 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 125-179. Recombinant protein control fragment (Product #RP-110133). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction