Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf146 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531672 Supplier Invitrogen™ Supplier No. PA531672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The C1orf146 gene, also known as FAM213A, encodes a protein involved in antioxidative and anti-inflammatory processes within the cell. Located on chromosome 1, FAM213A (family with sequence similarity 213 member A) has been implicated in several critical cellular functions, including the regulation of reactive oxygen species (ROS) and modulation of inflammatory responses. The protein encoded by C1orf146 is known to interact with peroxiredoxins, which are crucial for protecting cells from oxidative damage. Research has shown that alterations in FAM213A expression can affect cellular susceptibility to oxidative stress and may play a role in the pathogenesis of various diseases, including cancer and inflammatory disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf146
Applications Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene C1orf146
Gene Accession No. Q5VVC0
Gene Alias C1orf146; chromosome 1 open reading frame 146; RP11-163M2.4; uncharacterized protein C1orf146
Gene Symbols C1orf146
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C1orf146. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.