Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf149 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988915
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8915 Supplier Bioss Supplier No. BS9219R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The screening of cDNA expression libraries from human tumors with serum antibody (SEREX) has proven to be a powerful method for identifying the repertoire of tumor antigens recognized by the immune system of cancer patients, referred to as the cancer immunome. In this regard, cancer/testis (CT) antigens are of particular interest because of their immunogenicity and restricted expression patterns. Synovial sarcomas are striking with regard to CT antigen expression, however, highly expressed in sarcoma, CT antigens do not induce frequent humoral immune responses in sarcoma patients. Sera from two patients were used to immunoscreen cDNA libraries from two synovial sarcoma cell lines and normal testis, resulting in the identification of 113 distinct antigens.

Specifications

Antigen C1orf149
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MEAF6
Gene Accession No. Q9HAF1
Gene Alias 2310005N01Rik; 2810036M01Rik; BOS_3694; C1orf149; C3H1orf149; CDABP0189; CENP-28; centromere protein 28; Chromatin modification-related protein MEAF6; chromatin modification-related protein MEAF6-like; EAF6; Esa1-associated factor 6 homolog; Esa1p-associated factor 6 homolog; fj63g06; FLJ11730; meaf6; MYST/Esa1 associated factor 6; MYST/Esa1-associated factor 6; NY-SAR-91; Protein EAF6 homolog; RGD1310440; RP3-423B22.2; Sarcoma antigen NY-SAR-91; wu:fj63g06; zgc:100869
Gene Symbols MEAF6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf149.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64769
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.