Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf151 Monoclonal Antibody (OTI4H7)
GREENER_CHOICE

Catalog No. PIMA526031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526031 Supplier Invitrogen™ Supplier No. MA526031
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C1orf151 Monoclonal Antibody for Western Blot, IHC (P)

The exact function of C1orf151 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf151
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MINOS1
Gene Accession No. Q5TGZ0
Gene Alias 2310028O11Rik; C1orf151; MIC10; MICOS complex subunit MIC10; MICOS10; MINOS1; MIO10; mitochondrial contact site and cristae organizing system subunit 10; mitochondrial inner membrane organizing system 1; mitochondrial inner membrane organizing system protein 1; novel DUF543 domain containing protein; RGD1560187; RP5-1056L3.2; UPF0327 protein C1orf151; UPF0327 protein C1orf151 homolog
Gene Symbols MINOS1
Host Species Mouse
Immunogen Full length human recombinant protein of C1orf151 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440574
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.