Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf151 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116347
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116347 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116347 Supplier Invitrogen™ Supplier No. PA5116347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1orf151 Polyclonal Antibody for Western Blot

The exact function of C1orf151 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf151
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MINOS1
Gene Accession No. Q5TGZ0, Q7TNS2
Gene Alias 2310028O11Rik; C1orf151; MIC10; MICOS complex subunit MIC10; MICOS10; MINOS1; MIO10; mitochondrial contact site and cristae organizing system subunit 10; mitochondrial inner membrane organizing system 1; mitochondrial inner membrane organizing system protein 1; novel DUF543 domain containing protein; RGD1560187; RP5-1056L3.2; UPF0327 protein C1orf151; UPF0327 protein C1orf151 homolog
Gene Symbols MINOS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-33 amino acids from the N-terminal region of human MINOS1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 433771, 440574
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.