Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf163 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983385
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3385 Supplier Bioss Supplier No. BS15033R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for assembly of mitochondrial respiratory chain complex I and complex IV.

Specifications

Antigen C1orf163
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene COA7
Gene Accession No. Q96BR5
Gene Alias 2010305A19Rik; AI451324; Beta-lactamase hcp-like protein; C1orf163; Coa7; Cytochrome c oxidase assembly factor 7; cytochrome c oxidase assembly factor 7 (putative); D4Ertd796e; hcp beta-lactamase-like protein C1orf163; hcp beta-lactamase-like protein C1orf163 homolog; RESA1; respiratory chain assembly 1; Respiratory chain assembly factor 1; RGD1305274; Sel1 repeat containing 1; Sel1 repeat-containing protein 1; Selrc1
Gene Symbols COA7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf163.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65260
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.