Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf165 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563507 Supplier Invitrogen™ Supplier No. PA563507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLQLRHIKRP EGRKPSEVAH KSIEAVVARL EKQNGLSLGH STCPEEVFVE ASPGTEDMDS LEDAVVPRAL YEELLRNYQQ QQE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.

C1orf165, also known as CFAP299 (Cilia and Flagella Associated Protein 299), is located on chromosome 1 and encodes a protein that is implicated in ciliary function and structure. CFAP299 is essential for the proper assembly and motility of cilia and flagella, structures that are vital for various cellular processes, including cell motility, fluid movement, and signal transduction. Mutations in CFAP299 have been linked to primary ciliary dyskinesia (PCD), which is characterized by chronic respiratory tract infections, abnormal positioning of internal organs (situs inversus), and infertility due to defective cilia and flagella movement. The protein has been shown to interact with other ciliary proteins and play a role in the regulation of dynein arms and microtubule structures necessary for ciliary beating.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf165
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BEND5
Gene Accession No. Q7L4P6
Gene Alias 2310026E23Rik; BEN domain containing 5; BEN domain-containing protein 5; BEND5; C1orf165
Gene Symbols BEND5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf165. Recombinant protein control fragment (Product #RP-94362).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79656
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.