Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf165 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143071
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
C1orf165, also known as CFAP299 (Cilia and Flagella Associated Protein 299), is located on chromosome 1 and encodes a protein that is implicated in ciliary function and structure. CFAP299 is essential for the proper assembly and motility of cilia and flagella, structures that are vital for various cellular processes, including cell motility, fluid movement, and signal transduction. Mutations in CFAP299 have been linked to primary ciliary dyskinesia (PCD), which is characterized by chronic respiratory tract infections, abnormal positioning of internal organs (situs inversus), and infertility due to defective cilia and flagella movement. The protein has been shown to interact with other ciliary proteins and play a role in the regulation of dynein arms and microtubule structures necessary for ciliary beating.
Specifications
C1orf165 | |
Polyclonal | |
Unconjugated | |
BEND5 | |
2310026E23Rik; BEN domain containing 5; BEN domain-containing protein 5; BEND5; C1orf165 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
67621, 79656 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q7L4P6, Q8C6D4 | |
BEND5 | |
Peptide with sequence CKNEERREAKYNLQ (aa 408-421). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction