Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf172 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983388
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3388 Supplier Bioss Supplier No. BS15037R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plays a role in the regulation of the epidermis formation during early development. Required both as an inhibitor of basal cell proliferation and a promoter of differentiation of basal progenitor cell progeny.

Specifications

Antigen C1orf172
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KDF1
Gene Accession No. Q8NAX2
Gene Alias 1810019J16Rik; BOS_2376; C1orf172; C2H1orf172; chromosome 2 open reading frame, human C1orf172; Kdf1; Keratinocyte differentiation factor 1; LOC100357150; RGD1303271; RP11-344H11.3; uncharacterized LOC100357150; uncharacterized protein C1orf172 homolog
Gene Symbols KDF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf172.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126695
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.