Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1orf174 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | C1orf174 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170453
![]() |
Novus Biologicals
NBP170453 |
100 μL |
Each for $480.74
|
|
|||||
NBP17045320
![]() |
Novus Biologicals
NBP17045320UL |
20 μL | N/A | N/A | N/A | ||||
Description
C1orf174 Polyclonal specifically detects C1orf174 in Human samples. It is validated for Western Blot.Specifications
| C1orf174 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 339448 | |
| Synthetic peptides corresponding to C1ORF174 The peptide sequence was selected from the middle region of C1ORF174. Peptide sequence EAGVSVQQGAASLPLGGCRVVSDSRLAKTRDGLSVPKHSAGSGAEESNSS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| chromosome 1 open reading frame 174, RP13-531C17.2 | |
| C1ORF174 | |
| IgG | |
| 26 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title