Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf177 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113985 Supplier Invitrogen™ Supplier No. PA5113985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSMQKKKPRE LMNFKSFVEE LNSHHNKKHG VFSKLPRNPK TPTERIYWAN For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 79%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 79%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf177
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LEXM
Gene Accession No. Q3ZCV2
Gene Alias BC055111; C1orf177; cDNA sequence BC055111; LEM; LEXM; Lymphocyte expansion molecule; RGD1559493; similar to Hypothetical protein MGC58608; uncharacterized protein C1orf177; uncharacterized protein C1orf177 homolog
Gene Symbols LEXM
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C1orf177.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163747
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.