Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1ORF177 Rabbit anti-Mouse, Polyclonal, FabGennix

Catalog No. 502207594
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7594 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7594 Supplier Fabgennix Inc Supplier No. LEM101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1ORF177 Polyclonal antibody specifically detects C1ORF177 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen C1ORF177
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene LEXM
Gene Accession No. A2AVQ5
Gene Alias BC055111; C1orf177; cDNA sequence BC055111; LEM; LEXM; Lymphocyte expansion molecule; RGD1559493; similar to Hypothetical protein MGC58608; uncharacterized protein C1orf177; uncharacterized protein C1orf177 homolog
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 360-410 on mouse lyhmpocyte expansion molecule.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242602
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.