Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf180 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983393
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3393 Supplier Bioss Supplier No. BS15041R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AIDA, also known as C1orf80, is a 306 amino acid protein that belongs to the axin interactor family. Expressed in a variety of tissues, including skeletal muscle and heart, AIDA functions as a ventralizing factor during embryogenesis, disrupting Axin homodimerization and inhibiting Axin-mediated JNK activation. Axin, a scaffold protein, is important for both JNK signaling and the canonical Wnt pathway, two processes that play an essential role in embryonic dorsoventral patterning. Disruption of Axin by AIDA results in the negative regulation of JNK and Wnt signaling, thereby affecting embryonic developmental events. Three isoforms of AIDA exist due to alternative splicing events.

Specifications

Antigen C1orf180
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LINC01555
Gene Accession No. Q8NAE3
Gene Symbols LINC01555
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf180.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 439927
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.