Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf183 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3396 Supplier Bioss Supplier No. BS15044R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM212B (INKA2, Inka Box Actin Regulator 2) is a Protein Coding gene. An important paralog of this gene is ENSG00000284755. Inhibitor of the serine/threonine-protein kinase PAK4. Acts by binding PAK4 in a substrate-like manner, inhibiting the protein kinase activity.

Specifications

Antigen C1orf183
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM212B
Gene Accession No. Q9NTI7
Gene Alias 6530418L21Rik; BOS_3175; C1orf183; C3H1orf183; chromosome 3 open reading frame, human C1orf183; Fam212b; family with sequence similarity 212 member B; family with sequence similarity 212, member B; HInca-r; Induced in neural crest by AP2-alpha protein-related homolog; Inka box actin regulator 2; INKA2; LOC100351082; MInca-r; PAK4-inhibitor INKA2; protein FAM212B; RGD1306526; uncharacterized LOC100351082; uncharacterized protein C1orf183 homolog
Gene Symbols FAM212B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf183.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55924
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.