Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf186 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525856
Description
C1orf186 Polyclonal Antibody for Western Blot, IHC (P)
Enables erythropoietin receptor binding activity. Involved in erythropoietin-mediated signaling pathway and positive regulation of erythrocyte differentiation. Located in plasma membrane. RHEX (Regulator Of Hemoglobinization And Erythroid Cell Expansion) is a Protein Coding gene. Diseases associated with RHEX include Gallbladder Benign Neoplasm and Gallbladder Adenoma. Acts as a signaling transduction factor of the EPO-EPOR signaling pathway promoting erythroid cell differentiation.
Specifications
| C1orf186 | |
| Polyclonal | |
| Unconjugated | |
| C1orf186 | |
| C1orf186; chromosome 1 open reading frame 186; Regulator of hemoglobinization and erythroid cell expansion protein; regulator of human erythroid cell expansion; Regulator of human erythroid cell expansion protein; RHEX; uncharacterized protein C1orf186 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 440712 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6ZWK4 | |
| C1orf186 | |
| KLH conjugated synthetic peptide between 121-150 amino acids from the C-terminal region of human C1orf186. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction