Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf187 Rabbit anti-Bovine, Chicken, Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502206787
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6787 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6787 Supplier Fabgennix Inc Supplier No. DRAX101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1orf187 Polyclonal antibody specifically detects C1orf187 in Bovine, Chicken, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen C1orf187
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene DRAXIN
Gene Accession No. B6ZI38, P0C8S2, Q6PAL1, Q8NBI3
Gene Alias 2610109H07Rik; AGPA3119; C1orf187; Dorsal inhibitory axon guidance protein; dorsal repulsive axon guidance protein; Draxin; FLJ34999; MGC117222; Neucrin; neural tissue-specific cysteine-rich protein; PSEC0258; UNQ3119; UNQ3119/PRO10268
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 300-330 on C terminal region of Human Draxin protein.Region:NNKCFDDCMC VEGLRCYAKF HRNRRVTRRK
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374946, 419492, 506110, 691317, 70433, 736113
Target Species Bovine, Chicken, Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.