Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1ORF190 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983401
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3401 Supplier Bioss Supplier No. BS15049R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C1ORF190
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LURAP1
Gene Accession No. Q96LR2
Gene Alias C1orf190; Leucine repeat adapter protein 35A; leucine repeat adaptor protein 35a; Leucine rich adaptor protein 1; LRAP35A; LRP35A; LURAP1; NF-kappa-B activator C1orf190
Gene Symbols LURAP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1ORF190.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 541468
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.