Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf192 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983404
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3404 Supplier Bioss Supplier No. BS15051R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CFAP126 acts as a regulator of cilium basal body docking and positioning in mono- and multiciliated cells. Regulates basal body docking and cilia formation in multiciliated lung cells. Regulates kinocilium positioning and stereocilia bundle morphogenesis in the inner ear.

Specifications

Antigen C1orf192
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CFAP126
Gene Accession No. Q5VTH2
Gene Alias BOS_2643; C1orf192; C38H1orf192; C3H1orf192; C4H1orf192; CFAP126; chromosome 3 open reading frame, human C1orf192; chromosome 4 open reading frame, human C1orf192; cilia and flagella associated protein 126; Cilia- and flagella-associated protein 126; Flattop; FLTP; LOC100339157; Protein Flattop; RGD1562658; uncharacterized LOC100339157; UPF0740 protein C1orf192; UPF0740 protein C1orf192 homolog
Gene Symbols CFAP126
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf192.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257177
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.