Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555432 Supplier Invitrogen™ Supplier No. PA555432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKHVATVQNE EEAQKGKNYQ NGDVFGDEYR IKPVEEVKYM KNGAEEEQKI AARNQENLEK SASSNVRLKT NKEVPGLVHQ PRANMHISES QQEFFRMLDE KIEKGRDYCS EE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf21
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1orf21
Gene Accession No. Q9H246
Gene Alias C1orf21; Cell proliferation-inducing gene 13 protein; chromosome 1 open reading frame 21; PIG13; proliferation-inducing protein 13; uncharacterized protein C1orf21
Gene Symbols C1orf21
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf21. Recombinant protein control fragment (Product #RP-89715).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81563
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.