Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C1orf216 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565093
Description
Immunogen sequence: VQHLQVQERYK EQEKEKHHVH LVMYRRLALL QWIRGLQHQL IDQQARLQES FDTILDNRKE LIRCLQQRAA PSRP Highest antigen sequence identity to the following orthologs - mouse 89%, rat 88%.
C1orf216 is a protein coding gene.
Specifications
C1orf216 | |
Polyclonal | |
Unconjugated | |
C1orf216 | |
C1orf216; chromosome 1 open reading frame 216; UPF0500 protein; UPF0500 protein C1orf216 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127703 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TAB5 | |
C1orf216 | |
Recombinant Human C1orf216. Recombinant protein control fragment (Product #RP-104628). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction