Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf216 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983410
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3410 Supplier Bioss Supplier No. BS15058R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C1orf216 is a protein coding gene.

Specifications

Antigen C1orf216
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C1orf216
Gene Accession No. Q8TAB5
Gene Alias C1orf216; chromosome 1 open reading frame 216; UPF0500 protein; UPF0500 protein C1orf216
Gene Symbols C1orf216
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf216.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127703
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.