Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571024 Supplier Invitrogen™ Supplier No. PA571024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 77%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 77%; Rabbit: 79%; Rat: 86%.

ARMH1 (Armadillo repeat-containing protein 1) is a cytoplasmic protein that belongs to the armadillo repeat family of proteins. It is involved in the regulation of cell proliferation, differentiation, and apoptosis, and has been implicated in the development of various cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMH1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C1orf228
Gene Accession No. Q6PIY5
Gene Alias Armadillo-like helical domain containing protein 1; ARMH1; C1orf228; chromosome 1 open reading frame 228; NCRNA00082; p40; uncharacterized protein C1orf228
Gene Symbols C1orf228
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human C1orf228.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339541
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.