Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf25 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989345
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9345 Supplier Bioss Supplier No. BS9784R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that has some similarity to N2,N2-dimethylguanosine tRNA methyltransferase from other organisms. Studies of the mouse ortholog have shown that this protein plays a role in motor coordination and exploratory behavior, and it may also be involved in modulating postnatal neuronal functions. Alternatively spliced transcripts have been identified for this gene.

Specifications

Antigen C1orf25
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Trmt1l
Gene Accession No. Q7Z2T5
Gene Alias 1190005F20Rik; AW226554; bG120K12.3; C1orf25; MST070; MSTP070; TRM1 tRNA methyltransferase 1-like; TRM1L; Trm1-like; TRM1-like protein; TRMT1L; TRMT1-like protein; tRNA methyltransferase 1 homolog (S. cerevisiae)-like; tRNA methyltransferase 1 like; tRNA methyltransferase 1-like
Gene Symbols Trmt1l
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf25.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81627
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.