Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555016 Supplier Invitrogen™ Supplier No. PA555016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRILKTTEVP GFDKLVLIIP WVVMQELDRM KEGKLLKRAQ HKAIPAVHFI NDSLKNQDRK LWGQSIQLAS QKHYGLSDEN NDDRVLKCCL QHQE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 88%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf26
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SWT1
Gene Accession No. Q5T5J6
Gene Alias 1200016B10Rik; C1orf26; ca026; HsSwt1; SWT1; SWT1 RNA endoribonuclease homolog; SWT1 RNA endoribonuclease homolog (S. cerevisiae); SWT1, RNA endoribonuclease homolog; transcriptional protein SWT1
Gene Symbols SWT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf26. Recombinant protein control fragment (Product #RP-94083).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.