Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565094 Supplier Invitrogen™ Supplier No. PA565094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FCRTYDIHDPK SSARPADWKY QSGLSSSWLS LECTVHINIH IPLSATSVSY TLEKNTKNGL TRWAKEIENG VYLINGQVKD EDCDLLEGQK KSSRGNTQAT SHCFDVRVLT QLLLNSDHRS Highest antigen sequence identity to the following orthologs - mouse 88%, rat 85%.

May play a role in the trafficking of a subset of G-protein coupled receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf27
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ODR4
Gene Accession No. Q5SWX8
Gene Alias 1810053E15Rik; BC003331; C1orf27; cDNA sequence BC003331; chromosome 1 open reading frame 27; hODR-4; LAG1-interacting protein; mODR-4; odorant response abnormal 4; ODR4; odr-4; olfactory specific odr-4; protein odr-4 homolog; RGD1307830; similar to cDNA sequence BC003331; transactivated by recombinant transforming growth factor beta; transactivated by transforming growth factor beta protein 1; TTG1; TTG1A
Gene Symbols ODR4
Host Species Rabbit
Immunogen Recombinant Human C1orf27. Recombinant protein control fragment (Product #RP-105853).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54953
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.