Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf33 Mouse anti-Human, Polyclonal, Abnova™

Catalog No. 89333454 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-454 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-454 Supplier Abnova Corporation Supplier No. H00051154B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human C1orf33 protein.

This gene encodes a protein sharing a low level of sequence similarity with ribosomal protein P0. While the precise function of the encoded protein is currently unknown, it appears to be involved in mRNA turnover and ribosome assembly. [provided by RefSeq]

Specifications

Antigen C1orf33
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description mRNA turnover 4 homolog (S. cerevisiae)
Formulation PBS with no preservative; pH 7.4
Gene MRTO4
Gene Accession No. NM_016183
Gene Alias C1orf33, MRT4, dJ657E11.4
Gene Symbols MRTO4
Host Species Mouse
Immunogen C1orf33 (NP_057267, 1 a.a. ∽ 239 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51154
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.