Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf33 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983417
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3417 Supplier Bioss Supplier No. BS15064R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRTO4 is a protein sharing a low level of sequence similarity with ribosomal protein P0. While the precise function of the protein is currently unknown, it appears to be involved in mRNA turnover and ribosome assembly.

Specifications

Antigen C1orf33
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MRTO4
Gene Accession No. Q9UKD2
Gene Alias 2610012O22Rik; 60S acidic ribosomal protein PO; BOS_2333; C1orf33; dJ657E11.4; fb71a02; hypothetical protein LOC550350; LOC100353440; Mg684; mRNA turnover 4 homolog; mRNA turnover 4 homolog (S. cerevisiae); mRNA turnover protein 4 homolog; MRT4; MRT4 homolog, ribosome maturation factor; MRT4, mRNA turnover 4, homolog; MRT4, mRNA turnover 4, homolog (S. cerevisiae); Mrto4; MRTO4 ribosome maturation factor; muscle protein684; RGD1311709; Ribosome assembly factor Mrto4; wu:fb71a02; zgc:110388
Gene Symbols MRTO4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf33.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51154
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.