Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599222 Supplier Invitrogen™ Supplier No. PA599222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MMTAG2.

MMTAG2 gene ontology (GO) annotations include neutrophil degranulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf35
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C1orf35
Gene Accession No. Q99LX5, Q9BU76
Gene Alias 2310033P09Rik; C1orf35; chromosome 1 open reading frame 35; hMMTAG2; MMTAG2; multiple myeloma transforming 2; multiple myeloma tumor-associated protein 2; Multiple myeloma tumor-associated protein 2 homolog; RGD1304587; RIKEN cDNA 2310033P09 gene; similar to RIKEN cDNA 2310033P09
Gene Symbols 2310033P09Rik, C1orf35
Host Species Rabbit
Immunogen A synthesized peptide derived from human MMTAG2, corresponding to a region within C-terminal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67862, 79169
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.