Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf38 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983418
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3418 Supplier Bioss Supplier No. BS15065R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May constitute a control point in macrophage inflammatory response, promoting LPS-induced TNF production.

Specifications

Antigen C1orf38
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene THEMIS2
Gene Accession No. Q5TEJ8
Gene Alias basement membrane-induced; basement membrane-induced protein; C1orf38; ICB1; ICB-1; Induced by contact to basement membrane 1 protein; Protein ICB-1; Protein THEMIS2; RGD1561016; THEMIS2; thymocyte selection associated family member 2; Thymocyte-expressed molecule involved in selection protein 2
Gene Symbols THEMIS2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C1orf38.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9473
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.