Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1orf50 Antibody (OTI1C6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272188MFV450
Description
C1orf50 Monoclonal antibody specifically detects C1orf50 in Human, Mouse, Canine, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
C1orf50 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
79078 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
OTI1C6 | |
Flow Cytometry | |
chromosome 1 open reading frame 50, hypothetical protein LOC79078, MGC955 | |
Full length human recombinant protein of human (NP_077002) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Canine, Primate, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction