Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1orf50 Antibody (OTI1C6), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272188PEATT594
Description
C1orf50 Monoclonal specifically detects C1orf50 in Human, Mouse, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
C1orf50 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
C1orf50 | |
Full length recombinant protein of human C1orf50 (NP_077002) produced in HEK293T cells. | |
0.1 mL | |
79078.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1C6 | |
Flow Cytometry | |
chromosome 1 open reading frame 50, hypothetical protein LOC79078, MGC955 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Canine, Primate, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction