Learn More
Invitrogen™ C1orf50 Monoclonal Antibody (OTI1B11)

Description
C1orf50 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | C1orf50 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1B11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | C1orf50 |
| Gene Accession No. | Q9BV19 |
| Gene Alias | C15H1orf50; C1orf50; chromosome 1 open reading frame 50; chromosome 15 C1orf50 homolog; uncharacterized protein C1orf50; uncharacterized protein C1orf50 homolog |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.